missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRSK1 (SAD1) Recombinant Human Protein
BRSK1 (SAD1) is a serine/threonine kinase.
Marca: Invitrogen™ PR8037A
Detalles adicionales : Peso : 0.01000kg
Descripción
BRSK1 (SAD1) is a UV-induced DNA damage checkpoint kinase.
Especificaciones
NP_115806 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
BRSK1 (SAD1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
112.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |