Learn More
BrandTech™ BRAND™ 300μL Pipet Tips
Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene
Marca: BrandTech™ 702533
Detalles adicionales : Peso : 0.40000kg
Descripción
Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
- Pigment-free
- Available either in bulk packages or in convenient rack-and-stack formats
Especificaciones
5 to 300 μL | |
DiHEMDA- and oleamide-free | |
Bulk | |
Brand |
Non-sterile | |
For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility | |
1 bag of 1000 tips | |
Polypropylene |