Learn More
Invitrogen™ BRAF Recombinant Human Protein
BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus.
Marca: Invitrogen™ PV4172
Detalles adicionales : Peso : 0.00900kg
Descripción
BRAF is activated by somatic point mutation in human cancer, and is thought to play a role in the postsynaptic responses of hippocampal neuron.
Especificaciones
NP_004324.2 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
BRAF, BRAF1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
113.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
RAF Kinase Family | |
Recombinant | |
Baculovirus |