Learn More
Adaptador para llave de paso de vidrio borosilicatado Quickfit™ con patillas
Control flow rate. QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Marca: Quickfit MF14/1/SC
Detalles adicionales : Peso : 0.04000kg
Descripción
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Especificaciones
Stopcock Adapter | |
Borosilicate Glass | |
14/23 | |
Sockets |