Learn More
Bohlender™ Tapas de PTFE-EX del reactor BOLA™
Tapas de reactor fabricadas con PTFE-EX adecuadas para reactores de vidrio con brida plana Schott.
Marca:  Bohlender™ B170-16
Detalles adicionales : Peso : 5.06400kg
 Ver más versiones de este producto
Código de producto. 17106964
  • 638.12€ / cada uno
Fecha estimada de envío: 05-08-2024
para ver el stock
Descripción
Descripción
  • Made of static dissipative PTFE-EX
  • Suitable for glass reactors DN 100 with SCHOTT flat flange
  • With screw-in threads for connection of transition fittings
  • Bore with female thread M5x6 for earthing connection on the upper side of the lid
  • Easy assembly
  • Flexibly expandable
  • Compatible with glass reactors with SCHOTT-flat flange
  • Universal chemical resistance
  • With connectors for the use of existing equipment with ground joint or GL thread
Thread connections in detail:
  • Centric thread connection (purple) for insertion of a stirrer bearing
  • Vertical thread connection (blue) with parallel alignment to the stirrer shaft
  • Inclined thread connection (yellow) with direction straight to the stirrer shaft
  • Inclined thread connection (green) with direction aside the stirrer shaft
Especificaciones
Especificaciones
Cuello central de 12,7 mm (1/2 pulg), 2 cuellos laterales de 6,35 mm (1/4 pulg) y 3 de 12,7 mm (1/2 pulg).
PTFE-EX
NPT de 12,7 mm (1/2 pulg) directamente al eje, NPT de 12,7 mm (1/2 pulg) o NPT de 6,35 mm (1/4 pulg) a un lado del eje, NPT de 6,35 mm (1/4 pulg) vertical
6 roscas hembra NPT
Reactores de vidrio DN 100 con brida plana SCHOTT