missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLOC1S1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19687-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to NLOC1S1.
BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell′Angelica, 2004 ).Especificaciones
BLOC1S1 | |
Polyclonal | |
Unconjugated | |
BLOC1S1 | |
BLOC 1 subunit 1, BLOC1S1, BLOS1, GCN5 like protein 1, GCN5L1, MICoA, Protein RT14, RT14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14533, 2647, 288785 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZKU7, O55102, P78537 | |
Bloc1s1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |