missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Mouse Fc gamma RIIB / CD32b Protein, Avitag™,His Tag (SPR & BLI verified)

Biotinylated Protein

419.88€ - 1847.47€

Especificaciones

Número de acceso NP_001070657.1
Peso molecular 24.0 kDa
Fuente Biotinylated Mouse CD32b, Avitag,His Tag (SPR and BLI verified) (CDB-M82E8) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Accession # NP_001070657.1).
Inmunógeno Thr 40 - Arg 217
Requisitos de almacenamiento -20°C
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16477359
Ver documentos Detalles de la promoción
ACROBiosystems
CDB-M82E8-200UG
200 μg
1847.47€
200 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16487359
Ver documentos Detalles de la promoción
ACROBiosystems
CDB-M82E8-25UG
25 μg
419.88€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. Low affinity immunoglobulin gamma Fc region receptor II-b (FCGR2B) is also known as CD32b, FCG2, IGFR2. CD32B is expressed on B cells and myeloid dendritic cells. Ligation of CD32B on B cells downregulates antibody production and may, in some circumstances, promote apoptosis. Co-ligation of CD32B on dendritic cells inhibits maturation and blocks cell activation. CD32B may also be a target for monoclonal antibody therapy for malignancies.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 24.0 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

NP_001070657.1
Biotinylated Mouse CD32b, Avitag,His Tag (SPR and BLI verified) (CDB-M82E8) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Accession # NP_001070657.1).
-20°C
Fc gamma RIIB / CD32b
Mouse
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
24.0 kDa
Thr 40 - Arg 217
FCGR2B,CD32b,CD32,FCG2,IGFR2,CDw32
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
SDS
Documentos

Documentos

Product Certifications