Learn More
ACROBiosystems Biotinylated Human PD-L1 / B7-H1 Protein, Fc,Avitag™
MALS verified
Marca: ACROBiosystems PDL-H82F2-200UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 53.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP 054862.1 | |
53.6 kDa | |
200 µg | |
Phe 19- Arg 238 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Precision Avi | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>90% as determined by SDS-PAGE | |
Biotinylated Human PD-L1, Fc,Avitag (PDL-H82F2) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1). | |
-20°C | |
PD-L1 | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |