missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human NKG2C&CD94 Protein, His, Avitag
423.38€ - 1862.87€
Especificaciones
Peso molecular | 34.3 kDa |
---|---|
Fuente | Biotinylated Human NKG2C&CD94;, His,Avitag (NC4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94) (Accession # P26717-1 (NKG2C) & Q13241-1 (CD94)). |
Inmunógeno | Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94) |
Requisitos de almacenamiento | -20°C |
Alias de gen | NKG2C & CD94 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17206340
|
ACROBiosystems
NC4-H82E3-200ug |
200 μg |
1862.87€
200 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17216340
|
ACROBiosystems
NC4-H82E3-25ug |
25 μg |
423.38€
25 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. NKG2C/CD159c is used as a receptor for NK cells and some cytotoxic T cells to recognize MHC class I HLA-E molecules. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.3 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
34.3 kDa | |
Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94) | |
NKG2C & CD94 | |
Precision Avi | |
His Tag & Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
Biotinylated Human NKG2C&CD94;, His,Avitag (NC4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94) (Accession # P26717-1 (NKG2C) & Q13241-1 (CD94)). | |
-20°C | |
NKG2C & CD94 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |