Learn More
ACROBiosystems Biotinylated Human IL-2 R beta / CD122 Protein, His, Avitag™
Biotinylated Protein
399.17€ - 1733.51€
Especificaciones
Fuente Biotinylated Human IL-2 R beta, His,Avitag (ILB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Accession # P14784-1).
Nombre común IL-2 R beta
Sistema de expresión HEK293
Formato Powder
Conjugado Precision Avi
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16467749
Ver documentos Detalles de la promoción
ACROBiosystems
ILB-H82E3-200UG
200 µg
1733.51€
200 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 14-09-2022
para ver el stock
16477749
Ver documentos Detalles de la promoción
ACROBiosystems
ILB-H82E3-25UG
25 µg
399.17€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 14-09-2022
para ver el stock
Descripción
Descripción
Interleukin-2 receptor subunit beta (IL2RB) is also known as CD122, which Belongs to the type I cytokine receptor family. Type 4 subfamily. IL2RB contains 1 fibronectin type-III domain. IL2RB / CD122 is involved in T cell-mediated immune responses, is present in 3 forms with respect to ability to bind interleukin 2. The low affinity form is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from interleukin 2.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa. The protein migrates as 35-38 kDa and 42-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
Biotinylated Human IL-2 R beta, His,Avitag (ILB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Accession # P14784-1).
HEK293
Precision Avi
-20°C
IL-2 R beta
Powder
MABSol™ Biotin Labeled Proteins