Learn More
ACROBiosystems Biotinylated Human IL-13 Protein, His, Avitag™
Biotinylated Protein
Marca: ACROBiosystems IL3-H82E5-200ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Interleukin 13 (IL13) is also known as ALRH, BHR1and P600, is a single-chain glycosylated polypeptide, and is a cytokine critical in regulating inflammatory and immune responses. IL13 is secreted by many cell types, but especially by T helper type 2 (Th2) cells. IL-13 induces its effects through a multi-subunit receptor that includes the alpha chain of the IL-4 receptor (IL-4Rα) and at least one of two known IL-13-specific binding chains. The functions of IL-13 overlap considerably with those of IL-4, especially with regard to changes induced on hematopoietic cells, but these effects are probably less important given the more potent role of IL-4. IL-13 induces matrix metalloproteinases (MMPs) as part of a mechanism that protects against excessive allergic inflammation that predisposes to asphyxiation. IL-13 induces many features of allergic lung disease, including airway hyperresponsiveness, goblet cell metaplasia and mucus hypersecretion, which all contribute to airway obstruction.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 15.9 kDa. The protein migrates as 17 kDa and 28-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_002179.2 | |
15.9 kDa | |
200 µg | |
Gly 21 - Asn 132 | |
IL13,ALRH,BHR1,MGC116786,MGC116788,MGC116789,P600,Interleukin-13 | |
Precision Avi | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>90% as determined by SDS-PAGE | |
Biotinylated Human IL-13, His,Avitag (IL3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1). | |
-20°C | |
IL-13 | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |