missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human GITR / TNFRSF18 Protein, His,Avitag™

MALS verified

419.88€ - 1847.47€

Especificaciones

Número de acceso NP_004186.1
Peso molecular 18.1 kDa
Fuente Biotinylated Human GITR, His,Avitag™ (GIR-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).
Inmunógeno Gln 26 - Glu 161
Requisitos de almacenamiento -20°C
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17196331
Ver documentos Detalles de la promoción
ACROBiosystems
GIR-H82E1-200UG
200 μg
1847.47€
200 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17106341
Ver documentos Detalles de la promoción
ACROBiosystems
GIR-H82E1-25UG
25 μg
419.88€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Glucocorticoid-induced TNFR-related protein (GITR) is also known as Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), activation-inducible TNFR family receptor (AITR), CD antigen CD357, which is a member of the tumor necrosis factor receptor (TNF-R) superfamily. GITR is receptor for TNFSF18, which seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. GITR also mediated NF-kappa-B activation via the TRAF2/NIK pathway.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.1 kDa. The protein migrates as 22-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

NP_004186.1
Biotinylated Human GITR, His,Avitag™ (GIR-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).
-20°C
GITR
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
18.1 kDa
Gln 26 - Glu 161
AITR,GITR,TNFRSF18,CD357
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Videos
SDS
Documentos

Documentos

Product Certifications