Learn More
ACROBiosystems Biotinylated Human CD3 epsilon&CD3 delta Heterodimer Protein, His,Avitag™&Tag Free
MALS verified
Marca:  ACROBiosystems CDD-H82W6-25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17196211
  • 433.38€ / 25 microgramos
Fecha estimada de envío: 25-10-2022
para ver el stock
Descripción
Descripción
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Biotinylated Human CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 18.6 kDa (CD3E) and 13.4 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag and an Avi tag (Avitag™) at the
Especificaciones
Especificaciones
NP_000724.1 (CD3E) and NP_000723.1 (CD3D)
18.6 kDa (CD3E) and 13.4 kDa (CD3D)
25 µg
Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D)
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon
Precision Avi
PBS, pH7.4
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Biotinylated Human CD3E&CD3D; Heterodimer Protein, His,AviTag&Tag; Free (CDD-H82W6) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) and P04234-1 (CD3D)).
-20°C
CD3E and CD3D
His Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins