Learn More
ACROBiosystems Biotinylated Human CA125 / MUC16 (13810-14451) Protein, Fc, Avitag™
Biotinylated Protein
Marca:  ACROBiosystems CA5-H82F8-25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17196091
  • 399.17€ / 25 microgramos
Fecha estimada de envío: 14-10-2022
para ver el stock
Descripción
Descripción
The CA125, also known as the MUC16, is a mucin protein that may be found in type I transmembrane or secreted forms that are used monitor the progress of epithelial ovarian cancer therapy. The CA 125 molecule is almost certainly a glycoprotein with a predominance of O-linkages. It is heterogeneous with regard to both size and charge, most likely due to continuous deglycosylation of side chains during its life-span in bodily fluids. It exists as a very large complex (perhaps as much as 4 million daltons) under natural conditions.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 100.0 kDa. The protein migrates as 120-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_078966.2
100.0 kDa
25 µg
Pro 13810 - Pro 14451
CA125,CA-125,CA125MUC-16,FLJ14303,MUC16,mucin 16,mucin-16
Precision Avi
PBS, pH7.4
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Biotinylated Human CA125 (13810-14451), Fc,Avitag (CA5-H82F8) is expressed from human 293 cells (HEK293). It contains AA Pro 13810 - Pro 14451 (Accession # Q8WXI7-1).
-20°C
CA125
Fc Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins