Learn More
BD Red Nucleic Acid Stain
Useful for discrimination of viable from nonviable cells, DNA content analysis in fixed cells, or nuclear counterstaining for fixed cells in immunofluorescence or flow cytometry applications. BD Via-Probe™ Red Nucleic Acid Stain brightly stains nonviable or fixed cells with permeable membranes.
Marca: BD 565804
Detalles adicionales : Peso : 0.01000kg
Descripción
- Red Nucleic Acid Stain has an excitation maximum of 631nm, and is best excited by the red laser
- It has an emission maximum of 651nm
Especificaciones
-20°C | |
|
|
Red Nucleic Acid Stain |
0.5 mL | |
Red |