missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCS1L Mouse anti-Human, Mouse, Clone: 3G6H1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60212-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Five alternatively spliced transcripts encoding the same protein have been described.Especificaciones
BCS1L | |
Monoclonal | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CZP5, Q9Y276 | |
BCS1L | |
BCS1L Fusion Protein Ag18280 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
3G6H1 | |
Unconjugated | |
BCS1L | |
BCS1L, BJS, FLNMS, GRACILE, h BCS, h BCS1, Hs.6719, Mitochondrial chaperone BCS1, PTD | |
Mouse | |
Protein G | |
RUO | |
617, 66821 | |
-20°C | |
Liquid |