missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16417-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine, isoleucine, and valine. Multiple transcript variants encoding different isoforms have been found for this gene.Especificaciones
BCAT2 | |
Polyclonal | |
Unconjugated | |
BCAT2 | |
BCAT2, ECA40, Placental protein 18, PP18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12036, 587, 64203 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15382, O35854, O35855 | |
Bcat2 | |
BCAT2 Fusion Protein Ag9573 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |