missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAAT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15990-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a liver enzyme that catalyzes the transfer of C24 bile acids from the acyl-CoA thioester to either glycine or taurine, the second step in the formation of bile acid-amino acid conjugates. The bile acid conjugates then act as a detergent in the gastrointestinal tract, which enhances lipid and fat-soluble vitamin absorption. Defects in this gene are a cause of familial hypercholanemia (FHCA). Two transcript variants encoding the same protein have been found for this gene.Especificaciones
BAAT | |
Polyclonal | |
Unconjugated | |
BAAT | |
BAAT, BACAT, BAT, Glycine N choloyltransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
570 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14032 | |
BAAT | |
BAAT Fusion Protein Ag8860 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |