missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15280-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Subunit of the peripheral V1 complex of vacuolar ATPase essential for assembly or catalytic function. V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.Especificaciones
ATP6V1E1 | |
Polyclonal | |
Unconjugated | |
ATP6V1E1 | |
ATP6E, ATP6E2, ATP6V1E, ATP6V1E1, P31, V ATPase 31 kDa subunit, V ATPase subunit E 1, Vma4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11973, 297566, 529 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P36543, P50518, Q6PCU2 | |
ATP6V1E1 | |
ATP6V1E1 Fusion Protein Ag7399 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |