missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATIC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10726-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.Especificaciones
ATIC | |
Polyclonal | |
Unconjugated | |
ATIC | |
AICAR, AICARFT, ATIC, IMP synthase, IMPCHASE, Inosinicase, PURH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
471 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31939 | |
ATIC | |
ATIC Fusion Protein Ag1052 | |
150 μL | |
Primary | |
Human | |
Liquid |