Learn More
PIN Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610726
Detalles adicionales : Peso : 0.01000kg
Descripción
PIN (protein inhibitor of nNOS) is a small protein of 89 amino acids initially described as a light chain subunit of dynein and as an inhibitor of the neuronal nitric oxide synthase isoform (nNOS). In vitro, PIN has been reported to bind to a unique nNOS domain encompassing amino acids 163-245. PIN inhibits nNOS activity and blocks the formation of the active nNOS dimer. Although ubiquitously expressed, PIN levels are reportedly highest in brain and testis. Immunolocalization studies in Drosophila melanogaster detected a cytoplasmic distribution of PIN. Partial-loss-of-function of PIN resulted in morphologic and developmental changes in the bristles, wings, and female sterility of D. melanogaster. Deletion of PIN resulted in lethality with the characteristic morphology of apoptotic cells.
Immunofluorescence, Immunoprecipitation, Western Blotting
Especificaciones
PIN | |
Monoclonal | |
250μg/mL | |
Protein Inhibitor of nNOS | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot, Immunofluorescence | |
4 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Rat PIN aa. 1-89 | |
50 μg | |
Cell Biology | |
Rat, Human | |
IgG1 |
For Research Use Only.