Learn More
PIN Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
321.00€
Especificaciones
Antígeno | PIN |
---|---|
Clon | 4 |
Concentración | 250μg/mL |
Aplicaciones | Immunofluorescence, Western Blot |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15885609
|
BD Biosciences
610726 |
50 μg |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
PIN (protein inhibitor of nNOS) is a small protein of 89 amino acids initially described as a light chain subunit of dynein and as an inhibitor of the neuronal nitric oxide synthase isoform (nNOS). In vitro, PIN has been reported to bind to a unique nNOS domain encompassing amino acids 163-245. PIN inhibits nNOS activity and blocks the formation of the active nNOS dimer. Although ubiquitously expressed, PIN levels are reportedly highest in brain and testis. Immunolocalization studies in Drosophila melanogaster detected a cytoplasmic distribution of PIN. Partial-loss-of-function of PIN resulted in morphologic and developmental changes in the bristles, wings, and female sterility of D. melanogaster. Deletion of PIN resulted in lethality with the characteristic morphology of apoptotic cells.
Immunofluorescence, Immunoprecipitation, Western Blotting
Especificaciones
PIN | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Protein Inhibitor of nNOS |
4 | |
Immunofluorescence, Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
Rat PIN aa. 1-89 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.