Learn More
anti-IgG2a κ, Mouse, R-PE, Clone: G155-178, Isotype Control, BD
Marca: BD Biosciences 553457
Detalles adicionales : Peso : 0.99000kg
Descripción
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a κ
Species Reactivity: Non-human primate
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Intracellular Staining, Isotype Control
Especificaciones
Flow Cytometry, Western Blot | |
0.2mg/mL | |
Mouse | |
Affinity Purified | |
Undiluted at 4°C |
G155-178 | |
R-PE | |
TNP-keyhole limpet hemocyanin | |
0.1 mg | |
IgG2a κ |