missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ, Mouse, R-PE, Clone: G155-178, Isotype Control, BD

265.00€ - 494.00€

Especificaciones

Clon G155-178
Aplicaciones Flow Cytometry, Western Blot
Conjugado R-PE
Especie del huésped Mouse
Inmunógeno TNP-keyhole limpet hemocyanin
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
10226854
missing translation for 'viewDocuments' Promotion Details
BD Biosciences
553457
0.1 mg
494.00€
cada uno
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 15-04-2024
para ver el stock
Add to basket
 
15896648
missing translation for 'viewDocuments' Promotion Details
BD Biosciences
551438
50 Tests Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.

Host Species: Mouse
Clone: G155-178
Isotype: IgG2a κ
Species Reactivity: Non-human primate
Immunogen: TNP-keyhole limpet hemocyanin

Flow Cytometry, Intracellular Staining, Isotype Control

Specifications

Specifications

G155-178
R-PE
TNP-keyhole limpet hemocyanin
Affinity Purified
Flow Cytometry, Western Blot
Mouse
IgG2a κ
Undiluted at 4°C
Videos
SDS
Documents

Documents

Certificates