missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-1-Antichymotrypsin Mouse, Clone: AACT/1451 + AACT/1452, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 12-MSM3-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Alpha-1-Antichymotrypsin | |
Cytoplasmic | |
AACT/1451 + AACT/1452 | |
Unconjugated | |
NP_001076 | |
Mouse | |
65-76kDa | |
Protein A/G Purified | |
RUO | |
12 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
SERPINA3 | |
Recombinant human Antichymotrypsin (AACT) protein fragment (aa49-187) | |
HeLa Cells. Tonsil, Pancreas or Histiocytoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |