missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ALDH3B1 (Human) Recombinant Protein
Human ALDH3B1 full-length ORF ( AAH13584, 1 a.a. - 468 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000221-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MDPLGDTLRRLREAFHAGRTRPAEFRAAQLQGLGRFLQENKQLLHDALAQDLHKSAFESEVSEVAISQGEVTLALRNLRAWMKDERVPKNLATQLDSAFIRKEPFGLVLIIAPWNYPLNLTLVPLVGALAAGNCVVLKPSEISKNVEKILAEVLPQYVDQSCFAVVLGGPQETGQLLEHRFDYIFFTGSPRVGKIVMTAAAKHLTPVTLELGGKNPCYVDDNCDPQTVANRVAWFRYFNAGQTCVAPDYVLCSPEMQERLLPALQSTITRFYGDDPQSSPNLGRIINQKQFQRLRALLGCGRVAIGGQSDESDRYIAPTVLVDVQEMEPVMQEEIFGPILPIVNVQSLDEAIEFINRREKPLALYAFSNSSQVVKRVLTQTSSGGFCGNDGFMHMTLASLPFGGVGASGMGRYHGKFSFDTFSHHRACLLRSPGMEKLNALRYPPQSPRRLRMLLVAMEAQGCSCTLL
Especificaciones
AAH13584 | |
aldehyde dehydrogenase 3 family, member B1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDH3B1 | |
GST |
221 | |
Wheat germ expression system | |
10 μg | |
ALDH4, ALDH7, FLJ26433 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |