missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH3A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15090-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Aldh3A2 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh3A2 catalyzes the oxidation of long-chain aliphatic aldehydes to fatty acids. Mutations in the Aldh3A2 gene cause Sjogren-Larrson syndrome, an inherited neurocutaneous disorder. Patients with this disorder display ichthyosis, mental retardation and spastic diplegia. The pathogenesis of the cutaneous and neurological symptoms is thought to result from abnormal lipid accumulation in the membranes of skin and brain, the formation of aldehyde Schiff base adducts with amine-containing lipids or proteins, or defective eicosanoid metabolism.Especificaciones
ALDH3A2 | |
Polyclonal | |
Unconjugated | |
ALDH3A2 | |
Aldehyde dehydrogenase 10, ALDH10, ALDH3A2, FALDH, Fatty aldehyde dehydrogenase, SLS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11671, 224, 65183 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30839, P47740, P51648 | |
Aldh3a2 | |
ALDH3A2 Fusion Protein Ag1687 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |