missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27369-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ADAL is a putative nucleoside deaminase. It may catalyze the hydrolytic deamination of adenosine or some similar substrate and play a role in purine metabolism.Especificaciones
ADAL | |
Polyclonal | |
Unconjugated | |
ADAL | |
ADAL, adenosine deaminase like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
161823 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6DHV7 | |
ADAL | |
ADAL Fusion Protein Ag25949 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |