missing translation for 'onlineSavingsMsg'
Learn More

Cloruro de 4-clorobencidrilo, 98 %, Thermo Scientific Chemicals

Catalog Number p-7026663
Click to view available options
Cantidad:
25 g
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
134-83-8
C13H10Cl2
237.123
MFCD00000856
ALKWTKGPKKAZMN-UHFFFAOYSA-N
4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
241584
1-cloro-4-[cloro(fenil)metil]benceno
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
134-83-8
C13H10Cl2
237.123
MFCD00000856
ALKWTKGPKKAZMN-UHFFFAOYSA-N
4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
241584
1-cloro-4-[cloro(fenil)metil]benceno
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Chlorobenzhydryl chloride
Densidad 1.239
Punto de ebullición 159°C to 160°C (2 mmHg)
Temperatura de inflamación 194°C (381°F)
Índice de refracción 1.603
Cantidad 25 g
Número UN UN3265
Beilstein 2050236
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 237.13
Porcentaje de pureza 98%
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'