Learn More
3,5-Difluoro-2-metilbenzonitrilo, 97 %
55.86€ - 183.00€
Identificadores de productos químicos
CAS | 1003708-74-4 |
---|---|
Fórmula molecular | C8H5F2N |
Peso molecular (g/mol) | 153.132 |
Número MDL | MFCD09996934 |
Clave InChI | JHMDKULXRYFRBP-UHFFFAOYSA-N |
Sinónimo | 3,5-difluoro-2-methyl benzonitrile, pubchem8342 |
PubChem CID | 44887034 |
Nombre IUPAC | 3,5-difluoro-2-metilbenzonitrilo |
SMILES | CC1=C(C=C(C=C1C#N)F)F |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
1003708-74-4 | |
153.132 | |
JHMDKULXRYFRBP-UHFFFAOYSA-N | |
44887034 | |
CC1=C(C=C(C=C1C#N)F)F |
C8H5F2N | |
MFCD09996934 | |
3,5-difluoro-2-methyl benzonitrile, pubchem8342 | |
3,5-difluoro-2-metilbenzonitrilo |
Especificaciones
3,5-Difluoro-2-methylbenzonitrile | |
55°C to 57°C | |
C8H5F2N | |
UN3439 | |
JHMDKULXRYFRBP-UHFFFAOYSA-N | |
3,5-difluoro-2-metilbenzonitrilo | |
44887034 | |
97% |
1003708-74-4 | |
Blanco | |
MFCD09996934 | |
3,5-difluoro-2-methyl benzonitrile, pubchem8342 | |
CC1=C(C=C(C=C1C#N)F)F | |
153.132 | |
153.13 | |
Polvo cristalino |
Seguridad y manipulación
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes