missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de cloruro de 2-(dietilamino)etilo, +98 %, Thermo Scientific Chemicals

Catalog Number 11437417
Click to view available options
Cantidad:
100 g
500 g
2500 g
Este artículo no se puede devolver. Vea la política de devoluciones
869-24-9
C6H15Cl2N
172.09
MFCD00012519
RAGSWDIQBBZLLL-UHFFFAOYSA-N
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
13363
[H+].[Cl-].CCN(CC)CCCl
Este artículo no se puede devolver. Vea la política de devoluciones
869-24-9
C6H15Cl2N
172.09
MFCD00012519
RAGSWDIQBBZLLL-UHFFFAOYSA-N
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
13363
[H+].[Cl-].CCN(CC)CCCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-(Diethylamino)ethyl chloride hydrochloride
Punto de fusión 206°C to 210°C
pH 5
Fórmula lineal (CH3CH2)2NCH2CH2Cl·HCl
Cantidad 100 g
Número UN UN2928
Beilstein 3591991
Nombre IUPAC 2-cloro-N,N-dietiletanamina;clorhidrato
Formula Weight (peso de la fórmula) 172.1
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'