missing translation for 'onlineSavingsMsg'
Learn More

2,2-bis(4-Metoxifenil)etilamina, 98 %

Catalog Number 11430568
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
85336-83-0
C16H19NO2
257.333
MFCD04039769
VVTAUYUSSABNDB-UHFFFAOYSA-N
2,2-bis 4-methoxyphenyl ethylamine, 2,2-bis 4-methoxyphenyl ethanamine, 2,2-bis 4-methoxylphenyl ethylamine, benzeneethanamine,4-methoxy-b-4-methoxyphenyl
7016418
2,2-bis(4-metoxifenil)etanamina
COC1=CC=C(C=C1)C(CN)C2=CC=C(C=C2)OC
Este artículo no se puede devolver. Vea la política de devoluciones
85336-83-0
C16H19NO2
257.333
MFCD04039769
VVTAUYUSSABNDB-UHFFFAOYSA-N
2,2-bis 4-methoxyphenyl ethylamine, 2,2-bis 4-methoxyphenyl ethanamine, 2,2-bis 4-methoxylphenyl ethylamine, benzeneethanamine,4-methoxy-b-4-methoxyphenyl
7016418
2,2-bis(4-metoxifenil)etanamina
COC1=CC=C(C=C1)C(CN)C2=CC=C(C=C2)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,2-Bis(4-methoxyphenyl)ethylamine
Punto de fusión 45°C to 50°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 257.34
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'