Learn More
Hamilton™ 100 μL 800 Series Microliter Syringe
The 800 series Microliter syringes are the reinforced plunger version of the original 700 series syringes.
Marca:  Hamilton™ CAL7662-01
Detalles adicionales : Peso : 0.04600kg
 Ver más versiones de este producto
Código de producto. 17894342
  • 357.00€ / cada uno
Fecha estimada de envío: 04-07-2024
para ver el stock
Descripción
Descripción
  • Attached to the flange of the syringe is a blue Syringe Holder, which provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies.
  • Provides additional support to small-volume plungers that are prone to bending during injection.
  • This series of Microliter syringes is ideal for dispensing volumes up to 100 μL.
  • A variety of different terminations and needle options are available in this series.
Especificaciones
Especificaciones
Autoclavable
No Needle Included
0.305 in.
7.75 mm
Microliter Syringe
115°C
810 RN
1
No Needle Included
Removable
Stainless Steel
0.061 in.
1.46 mm
No Needle Included
239°F
50°F
10°C
100 μL
No Needle Included