Learn More
Hamilton™ 100 μL 800 Series Microliter Syringe
Descripción
- Attached to the flange of the syringe is a blue Syringe Holder, which provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies.
- Provides additional support to small-volume plungers that are prone to bending during injection.
- This series of Microliter syringes is ideal for dispensing volumes up to 100 μL.
- A variety of different terminations and needle options are available in this series.
Especificaciones
Especificaciones
| Esterilizables en autoclave | Not Autoclavable |
| Material | Stainless Steel |
| Calibre de aguja | 22s G |
| Diámetro interior (imperial) | 0,061 in |
| Diámetro interior de la aguja (imperial) | 0,0059 in |
| Diámetro interior de la aguja (métrico) | 0.15 mm |
| Diámetro exterior de la aguja (imperial) | 0,0283 in |
| Diámetro exterior (imperial) | 0,305 in |
| Diámetro interior (métrico) | 1.46 mm |
| Diámetro exterior (métrico) | 7.75 mm |
| Mostrar más |