Learn More
Hamilton™ 10 μL 900 Series Microliter Syringe
The 900 series syringe is the economical version of the 800 series syringe.
Marca:  Hamilton™ CAL80387
Detalles adicionales : Peso : 0.02300kg
 Ver más versiones de este producto
Código de producto. 17884332
  • 256.00€ / cada uno
Fecha estimada de envío: 05-07-2024
para ver el stock
Descripción
Descripción
  • This series of Microliter syringes is ideal for dispensing volumes up to 10 μL.
  • The syringe holder is permanently attached to the syringe barrel, reducing the initial cost of the syringe but eliminating the possibility of replacement components.
Especificaciones
Especificaciones
Autoclavable
23s G
0.005 in.
0.0187 in.
0.485 mm
0.47 mm
Microliter
115°C
901 N
10 μL
Cone
1
Stainless Steel
0.019 in.
0.13 mm
0.260 in.
6.60 mm
1.71 in.
239°F
50°F
10°C
43.4 mm
Cemented
Microliter Syringe