Learn More
Hamilton™ 10 μL 900 Series Microliter Syringe
The 900 series syringe is the economical version of the 800 series syringe.
Marca: Hamilton™ CAL7659-01
Detalles adicionales : Peso : 0.04500kg
Descripción
- This series of Microliter syringes is ideal for dispensing volumes up to 10 μL.
- The syringe holder is permanently attached to the syringe barrel, reducing the initial cost of the syringe but eliminating the possibility of replacement components.
Especificaciones
Autoclavable | |
No Needle Included | |
0.260 in. | |
6.60 mm | |
Microliter Syringe | |
115°C | |
901 RN | |
1 | |
No Needle Included | |
Removable |
Borosilicate Glass | |
0.019 in. | |
0.485 mm | |
No Needle Included | |
239°F | |
50°F | |
10°C | |
10 μL | |
No Needle Included |