missing translation for 'onlineSavingsMsg'
                
Learn More
Learn More
Chromogranin A Antibody (CL0166) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-52869IR
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                Este artículo no se puede devolver.
                                Vea la política de devoluciones
                                
                        
                    
                Descripción
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
            Especificaciones
| Chromogranin A | |
| Monoclonal | |
| DyLight 755 | |
| betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I | |
| This antibody was developed using a recombinant protein derived from P10645, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers | |
| 1113 | |
| Store at 4⪚C in the dark. | |
| IgG1 | 
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0166 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Corrección del contenido de un producto
                
                Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
			Chromogranin A Antibody (CL0166) [DyLight 755], Novus Biologicals™ > 0.1 mL; DyLight 755
            ¿Detecta una oportunidad de mejora?Comparta una corrección de contenido