missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF207 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14814-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ZNF207 is a new candidate transcription factor.Especificaciones
ZNF207 | |
Polyclonal | |
Unconjugated | |
ZNF207 | |
zinc finger protein 207, ZNF207 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22680, 303763, 7756 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43670, Q9JMD0 | |
Zfp207, ZNF207 | |
ZNF207 Fusion Protein Ag6564 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |