missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, ACS Reagent, 98 to 101%, Honeywell Fluka™

Catalog Number 15615290
Click to view available options
Cantidad:
500 g
2.5 kg
12 kg
Envase:
Drum with Inliner
Poly Bottle
Este artículo no se puede devolver. Vea la política de devoluciones
5970-45-6
C4H10O6Zn
219.498
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
Este artículo no se puede devolver. Vea la política de devoluciones
5970-45-6
C4H10O6Zn
219.498
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Zinc acetate dihydrate
Punto de fusión ∼100°C
Color Gray
pH 6 to 8 (at 20°C)
Cantidad 12 kg
Fórmula lineal Zn(CH3COO)2 · 2H2O
Número UN UN3077
Beilstein 3732513
Información de solubilidad In water 311g/L at 20°C
Formula Weight (peso de la fórmula) 219.51g/mol
Porcentaje de pureza 98.0 to 101.0%
Olor Vinegar-like
Envase Drum with Inliner
Aspecto Crystalline
Densidad 1.750g/cm3 at 20°C
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'