missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNT10A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26238-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is strongly expressed in the cell lines of promyelocytic leukemia and Burkitt′s lymphoma. In addition, it and another family member, the WNT6 gene, are strongly coexpressed in colorectal cancer cell lines. The gene overexpression may play key roles in carcinogenesis through activation of the WNT-beta-catenin-TCF signaling pathway. This gene and the WNT6 gene are clustered in the chromosome 2q35 region.Especificaciones
WNT10A | |
Polyclonal | |
Unconjugated | |
WNT10A | |
Protein Wnt 10a, WNT10A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22409, 80326 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70701, Q9GZT5 | |
WNT10A | |
WNT10A Fusion Protein Ag23963 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |