Learn More
Medicom™ WeePro Type PB 6 Overshoe with Anti-slip Sole
Click to view available options
WeePro Type PB 6 Overshoe with Anti-slip Sole; Size: Large/X-Large
Descripción
- Lint-free material to avoid any contamination in a controlled critical environment
- Thick PVC sole with a high anti-slip property over dry or wet floors
- Elastic wrap at ankle and heel to keep the overshoes straight
- Anti-static material to allow easy dissipation of electrostatic charges
- Ideal protection against low concentration chemical splashes, powder, adhesives, fiberglass, solvents, paints
Especificaciones
Tamaño | Large/X-Large |
Material | Microporous Fabric |
Latex Free | No |
Color | White |
Desechables | Single-use |
Esterilidad | Non-sterile |
Tipo de tapón | Elastic Ankle |
Tipo de suela | Anti-slip |
Resistencia | Light Chemical Splash |
Tipo de producto | Controlled Environments Shoe Covers |