missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWF/VWFpp Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11778-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody raised against the N-terminal region of pre-pro-vWF recognizes von Willebrand antigen 2 (75-100 kDa) and pro-vWF (309-320 kDa).
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.Especificaciones
VWF/VWFpp | |
Polyclonal | |
Unconjugated | |
VWF | |
F8VWF, von Willebrand antigen II, von Willebrand factor, VWD, VWF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116669, 22371, 7450 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04275, Q62935, Q8CIZ8 | |
VWF | |
VWF, VWFpp Fusion Protein Ag2386 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |