missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamina B12, + 98 % (base de peso seco), Thermo Scientific Chemicals
Click to view available options
Cantidad:
1 g
5 g
25 g
Descripción
Vitamin B12 is used as a nutritional supplement and stabilizer in foods. It is also used as a medicine and animal feed supplement. It plays an important role in the synthesis of myelin. It is also used as an antipernicious anemia factor. Further, it is used to treat cyanide poisoning and hereditary deficiency of transcobalamin II.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Vitamin B12 |
Envase | Frasco de vidrio |
Cantidad | 1 g |
Beilstein | 4122889 |
Sensibilidad | Hygroscopic |
Información de solubilidad | Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in chloroform and acetone. |
Formula Weight (peso de la fórmula) | 1355.38 |
Porcentaje de pureza | ≥98% |
Forma física | Polvo cristalino |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'