missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Monoclonal Antibody (MIH64), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen M035T03Y07
201.67 EUR hasta el 2023-03-27
Use el código promocional "20585" para aplicar la oferta.
Detalles adicionales : Peso : 0.02000kg
Descripción
VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow CytometryEspecificaciones
VISTA | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9D659 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
MIH64 | |
NovaFluor Yellow 730 | |
VSIR | |
4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
Affinity chromatography | |
RUO | |
74048 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |