Learn More
Novus Biologicals™ Recombinant Mouse Apolipoprotein A-I/ApoA1 His Protein
Tienda Bio Techne Productos
Click to view available options
Cantidad:
0,1 mg
0,5 mg
Tamaño de la unidad:
0.10 miligramo
0.50 miligramo
Descripción
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-264 of Mouse APOA1 The Recombinant Mouse Apolipoprotein A-I/ApoA1 Protein is derived from E. coli. The Recombinant Mouse Apolipoprotein A-I/ApoA1 Protein has been validated for the following applications: SDS-Page.
Especificaciones
Especificaciones
| Concentración | 1mg/mL |
| Para utilizar con (aplicación) | SDS-PAGE |
| Formulación | Phosphate buffered saline (pH7.4) containing 20% glycerol, 1mM DTT |
| ID de gen (Entrez) | 128240 |
| Peso molecular | TMW: 30.3kDa |
| Cantidad | 0,1 mg |
| Requisitos de almacenamiento | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Alias de gen | AIBPAI-BP, apolipoprotein A-I binding protein, apolipoprotein A-I-binding protein, MGC119143, MGC119144, MGC119145, YjeF N-terminal domain-containing protein 1, YjeF_N1, YJEFN1apoA-I binding protein |
| Nombre común | APOA1 |
| Conjugado | Unconjugated |
| Mostrar más |
For research use only and is not approved for use in humans or in clinical diagnosis