Learn More
VGLUT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55491-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.Especificaciones
VGLUT1 | |
Polyclonal | |
Unconjugated | |
SLC17A7 | |
BNPI, SLC17A7, VGLUT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57030 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9P2U7 | |
SLC17A7 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |