missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14345-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by VBP1 interacts with the Von Hippel-Lindau protein to form an intracellular complex. The encoded protein functions as a chaperone protein, and may play a role in the transport of the Von Hippel-Lindau protein from the perinuclear granules to the nucleus or cytoplasm. Alternative splicing and the use of alternate transcription start sites results in multiple transcript variants encoding different protein isoforms.Especificaciones
VBP1 | |
Polyclonal | |
Unconjugated | |
VBP1 | |
HIBBJ46, PFD3, PFDN3, Prefoldin subunit 3, VBP 1, VBP1, VHL binding protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22327, 681825, 7411 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61758, P61759 | |
VBP1 | |
VBP1 Fusion Protein Ag5685 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |