Learn More
URAT1 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14937-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody recognizes the native and glycosylated URAT1 between 65 and 70 kDa as well as the truncated 40 kDa protein.
SLC22A12 is required for efficient urate re-absorption in the kidney. SLC22A12 regulates blood urate levels. SLC22A12 mediates saturable urate uptake by facilitating the exchange of urate against organic anions.The protein encoded by this gene is a urate transporter and urate-anion exchanger which regulates the level of urate in the blood. This protein is an integral membrane protein primarily found in kidney. Two transcript variants encoding different isoforms have been found for this gene.Especificaciones
URAT1 | |
Polyclonal | |
Unconjugated | |
SLC22A12 | |
OAT4L, OATL4, Renal specific transporter, RST, SLC22A12, URAT1, Urate anion exchanger 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100520540, 116085, 20521 | |
-20°C | |
Liquid |
Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CFZ5, Q96S37 | |
SLC22A12 | |
URAT1 Fusion Protein Ag6770 | |
20 μL | |
Primary | |
Porcine, Human, Mouse | |
Antibody | |
IgG |