missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28114-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.Especificaciones
UQCC | |
Polyclonal | |
Unconjugated | |
UQCC1 | |
BFZB, BZFB, C20orf44, CBP3, UQCC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55245 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NVA1 | |
UQCC1 | |
UQCC Fusion Protein Ag27123 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |