missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWIST2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11752-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Binds to the E-box consensus sequence 5′-CANNTG-3′ as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.Especificaciones
TWIST2 | |
Polyclonal | |
Unconjugated | |
TWIST2 | |
bHLHa39, Dermis expressed protein 1, Dermo 1, DERMO1, twist homolog 2 (Drosophila), Twist related protein 2, TWIST2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117581, 13345 | |
-20°C | |
Liquid |
Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WVJ9, Q9D030 | |
Twist2 | |
TWIST2 Fusion Protein Ag2348 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |